missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide solution, 1M EU Pharmacopoeia, Fisher Chemical™ Product Code.: 12910894
compliance-icons

Sodium Hydroxide solution, 1M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12910894
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12910894

Brand: Fisher Chemical VL 053 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 1310-73-2, 7732-18-5
Packaging Bottle
Quantity 1000 mL
Molecular Formula HNaO
UN Number 1824
Concentration 1M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
PubChem CID 14798
ChEBI CHEBI:32145
Grade Eu. Ph. Reference
Chemical Name or Material Sodium hydroxide solution
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1101751 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberVL 053 1000
Lot Number1099614 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberVL 053 1000
Lot Number1080784 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 053 1000
Lot Number1070446 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 053 1000
Lot Number1032632 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 053 1000
compliance-icons
Product Identifier
  • Sodium hydroxide 1M. Volumetric Solution European Pharmacopoeia Ref: 3006300
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide solution, 1M EU Pharmacopoeia, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.