missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate anhydrous, For ACS analysis, 99.32%, MP Biomedicals™ Product Code.: 11474662

Sodium sulfate anhydrous, For ACS analysis, 99.32%, MP Biomedicals™

Product Code. 11474662 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11474662

Brand: MP Biomedicals 0219144401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 0.9932
Formula Weight 142.04
Density 2.7g/cm3 at 20°C
Chemical Name or Material Sodium sulfate anhydrous, For ACS analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate anhydrous, For ACS analysis, 99.32%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.