Learn More
MP Biomedicals Sodium Test Diet, Powder
Low sodium salt mixture.
810.00 EUR - 3055.00 EUR
Specifications
Product Type | Animal Diet |
---|---|
Autoclavable | Autoclavable |
Disposable | Single-use |
Format | Powder |
Sterility | Non-sterile |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11437101
|
MP Biomedicals
0296023310 |
10 kg |
810.00 EUR
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447101
|
MP Biomedicals
0296023320 |
20 kg |
1405.00 EUR
20kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457101
|
MP Biomedicals
0296023350 |
50 kg |
3055.00 EUR
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Low sodium salt mixture
- Contains minerals and vitamin mix, choline, casein, ethoxyquin and other nutritious items
Specifications
Animal Diet | |
Single-use | |
Non-sterile | |
1 ft. |
Autoclavable | |
Powder | |
Room Temperature | |
0.3048 m |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
MP Biomedicals Sodium Test Diet, Powder