missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium valproate, 99.9%, MP Biomedicals™
73.70 EUR - 392.00 EUR
Chemical Identifiers
CAS | 1069-66-5 |
---|---|
Molecular Formula | C8H15NaO2 |
Molecular Weight (g/mol) | 166.196 |
InChI Key | AEQFSUDEHCCHBT-UHFFFAOYSA-M |
Synonym | sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin |
PubChem CID | 16760703 |
ChEBI | CHEBI:9925 |
IUPAC Name | sodium;2-propylpentanoate |
SMILES | CCCC(CCC)C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474361
|
MP Biomedicals
0215206410 |
10 g |
73.70 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484361
|
MP Biomedicals
0215206425 |
25 g |
116.00 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494361
|
MP Biomedicals
0215206480 |
100 g |
392.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
1069-66-5 | |
166.196 | |
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin | |
CHEBI:9925 | |
CCCC(CCC)C(=O)[O-].[Na+] |
C8H15NaO2 | |
AEQFSUDEHCCHBT-UHFFFAOYSA-M | |
16760703 | |
sodium;2-propylpentanoate |
Specifications
1069-66-5 | |
Crystalline Powder | |
C8H15NaO2 | |
AEQFSUDEHCCHBT-UHFFFAOYSA-M | |
sodium;2-propylpentanoate | |
16760703 | |
166.2 |
White | |
0.999 | |
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin | |
CCCC(CCC)C(=O)[O-].[Na+] | |
166.196 | |
CHEBI:9925 | |
Sodium valproate |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium Valproate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Valproate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title