Learn More
Hamilton™ SoftGrip™ Adjustable Volume Multiple Channel Pipette
With a dual indicator that improves the volume adjustment accuracy by removing parallax errors.
Brand: Hamilton™ 55019-52
This item is not returnable.
View return policy
Description
- Wide range of volumes available
- 8 or 12 channels
- Unique, long tip ejector lever and movable lower body can be rotated a full 360° relative to handle
- Isolated volume adjustment so there is no need for a locking mechanism to prevent accidental volume adjustment
- Universal calibration key that reduces the need for costly calibration services
- Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
0.50±% | |
Autoclavable | |
8 | |
Multi-Channel Adjustable Volume Pipette | |
0.30±% |
8 channels | |
Brick Red | |
SoftGrip | |
30 to 300 μL | |
1/Pk. |
Product Content Correction
Product Title
Hamilton™ SoftGrip™ Adjustable Volume Multiple Channel Pipette > Volume: 30-300uL; 8 channel
Spot an opportunity for improvement?Share a Content Correction