Learn More
Cole-Parmer™ Soil Auger Kit with Carrying Case
Soil Auger Kit with Carrying Case - Obtain 3 in. diameter samples in nearly all soil conditions with this complete auger kit.
Brand: Cole-Parmer™ 99026-40
This item is not returnable.
View return policy
Description
- Nickel-plated carbon steel material protects against rust and wear
Specifications
Nickel-plated | |
Soil Auger Kit | |
3 in. general-purpose auger, 3 in. clay auger, 3 in. sand auger, 3 in. mud auger, handgrip, two 45 in. auger extensions, two tools to dissemble extensions, carrying case |
Product Content Correction
Product Title
Cole-Parmer™ Soil Auger Kit with Carrying Case > Soil Auger Kit with Carrying Case; Material: Nickel-Plated; Sample Depth: 3 in.
Spot an opportunity for improvement?Share a Content Correction