Documents
Scientific Resources
Application Notes
- 1
- 2
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Brand: Thermo Scientific™ 60309-004
301.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
10mg/2mL 96 well plate format
Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
SOLA products are also available in cartridge format.
10 mg/2 mL; SOLA WCX 96 well SPE plate; 1 Pk. | |
Bioanalytical Applications - Strong Ion-Exchange Retention of Strong Basic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds | |
Mixed Mode Weak Cation Exchanger, Polymeric | |
2 mL |
10 mg | |
SOLA | |
96-well SPE Plate |