Learn More
uvex Sport Hygiene White PU 65808 Low Shoe - Width 11
Modern, metal-free, ultra-lightweight and flexible S2 safety slip on shoe with removable hook and loop strap for optional colored marking
119.00 EUR
Specifications
Color | White |
---|---|
Material | Microfiber |
Toe Material | Composite |
Disposable | Reusable |
Certifications/Compliance (Europe) | EN ISO 20345:2011 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17202351
|
uvex
6580835 |
35 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17212351
|
uvex
6580836 |
36 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17222351
|
uvex
6580837 |
37 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17232351
|
uvex
6580838 |
38 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17242351
|
uvex
6580839 |
39 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17252351
|
uvex
6580840 |
40 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17262351
|
uvex
6580841 |
41 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17272351
|
uvex
6580842 |
42 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17282351
|
uvex
6580843 |
43 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17292351
|
uvex
6580844 |
44 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17202361
|
uvex
6580845 |
45 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17212361
|
uvex
6580846 |
46 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17222361
|
uvex
6580847 |
47 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17232361
|
uvex
6580848 |
48 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17242361
|
uvex
6580849 |
49 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17252361
|
uvex
6580850 |
50 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17262361
|
uvex
6580851 |
51 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
17272361
|
uvex
6580852 |
52 |
119.00 EUR
1 pair |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
Description
Uvex 1 Sport Hygeine Shoe 6580.8 S2 SRC
- Upper made from water-repellent microfiber material that is easy to clean
- Minimum number of seams to reduce contamination
- Made from synthetic material, suitable for people allergic to chrome
- Washable up to 30°C
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume
- Resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with good shock, absorption and excellent slip resistance
- Breathable materials
- Virtually seam-free construction to eliminate pressure points
- Removable anti-static comfort insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6580.8
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 35 to 52
Specifications
White | |
Composite | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S2 | |
ESD Safety Shoe |
Microfiber | |
Reusable | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Sport Hygiene White PU 65808 Low Shoe - Width 11