Learn More
Usbeck Laborgeraete™ Stainless Steel Alcohol Burners
Brand:  Usbeck Laborgeraete™ 1500
 View more versions of this product
Product Code. 10251031
  • 23.55 EUR / Each
Estimated Shipment: 05-06-2025
Specifications
Spirit Burner
60 mL
1 Ea.
Stainless Steel
Adjustable Wick and Cap
75 g
Documents
One moment while we fetch your results.
Product Title
Usbeck Laborgeraete™ Stainless Steel Alcohol Burners > Capacity: 60mL; Adjustable Wick and Cap