Learn More
Bochem™ Cork Bore Set with Ejector
Brand:  Bochem™ 12431
 View more versions of this product
Product Code. 10396381
  • 217.00 EUR / Each
Estimated Shipment: 30-04-2025
Specifications
Cork Borer Set
Tube: Brass, nickel plated; Cork borer: Brass chromed; Ejector: Stainless steel w/ plastic knob
6Pack
Cork Borer Set
Sizes 8.75, 10.0, 11.25
Product Title
Bochem™ Cork Bore Set with Ejector > Sizes: 8.75, 10.0, 11.25