Learn More
Contec™ Stainless Steel Multi-Bucket System
Stainless Steel Multi-Bucket System offers maximum flexibility and can be used with two or three 7 gallon (27 L) buckets depending upon your in-house protocols.
Brand: Contec™ 2707
1879.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lightweight, stainless steel, and easy to maneuver multi-bucket system offers maximum flexibility and can be used with two or three 7- gallon (27 L) buckets depending upon in-house protocols.
- An extendable frame pulls out of the front of the fully autoclavable cart to accommodate an additional bucket, part number 2617.
- The cart features non-marring run quiet wheels.
- Buckets are deep drawn and seamless reducing the risk of microbial contamination
- The removable handle makes it convenient for autoclaving and storage.
- A lip around the top edge of the buckets allows for easy lifting.
- Autoclavable at 121°C for 30 minutes
- 2700 multi-bucket system includes heavy-duty stainless steel downpress wringer that is compatible with all sizes of VertiKlean™, VertiKlean™ MAX™, QuickTask™, and Edgeless™ Mops.
- 2707 multi-bucket system offers the ability to customize the system with whichever wringer best coincides with in-house protocols. The system is compatible with two stainless steel downpress wringer options to accommodate both flat and string mops
Specifications
Multiple Bucket System | |
Autoclavable | |
Ideal for use in life sciences, microelectronics, semiconductor or any critical manufacturing environment | |
Non-sterile |
Stainless Steel | |
Reusable | |
Two 7 gal. Buckets, cart with casters (wringer options sold separately) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Contec™ Stainless Steel Multi-Bucket System > Includes: Two 7 gal. buckets, cart with casters
Spot an opportunity for improvement?Share a Content Correction