Learn More
Bochem™ Stainless Steel Pinch Cock
Brand:  Bochem™ 12545
Product Code. 10799271
  • 14.75 EUR / Each
Estimated Shipment: 27-05-2025
Specifications
Hoffman Pinchcock
Stainless Steel
40 mm
Product Title
Bochem™ Stainless Steel Pinch Cock > Length: 40mm