Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Brand: RETSCH 03.729.0039
This item is not returnable.
View return policy
Specifications
Push-fit Feed Chute | |
For Holder 15/40 | |
Stainless Steel |
210 x 36 mm | |
210 mm | |
36 mm |
Product Content Correction
Product Title
RETSCH Stainless Steel Push-fit Feed Chutes > Dimensions (L x W): 210 x 36mm
Spot an opportunity for improvement?Share a Content Correction