Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Brand: Kinematica™ 35095002
This item is not returnable.
View return policy
Specifications
Sieve | |
PX MFC 90 D | |
Stainless Steel | |
0.2 mm |
Product Content Correction
Product Title
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill > Sieve Number: 0.2mm
Spot an opportunity for improvement?Share a Content Correction