Learn More
Bel-Art™ Stainless Steel Spoons
Double-ended with two bowl sizes.
73.00 EUR - 157.99 EUR
Specifications
Product Type | Spoon |
---|---|
Material | Stainless Steel |
Autoclavable | Autoclavable |
Style | Solid, Rounded Shaft |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Finish | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Finish | Price | Quantity & Availability | |||||
15550122
|
Bel-Art™
H36729-0018 |
0.8 mL small spoon, 2.8 mL large spoon | 18 cm | Plain-polished 304 grade stainless-steel |
73.00 EUR
Pack of 2 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
15570122
|
Bel-Art™
H36729-0025 |
1.8 mL small spoon, 5.0 mL large spoon | 25 cm | Plain-polished 304 grade stainless-steel |
93.40 EUR
Pack of 2 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
11726545
|
Bel-Art™
F36730-0030 |
2.0 mL small spoon, 8.0 mL large spoon | 30 cm | Fluo-Kem Teflon FEP coated ends |
157.99 EUR
Pack of 2 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
11706545
|
Bel-Art™
F36730-0015 |
0.5, 2 mL | 15 cm | Fluo-Kem Teflon FEP coated |
143.00 EUR
Pack of 2 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
11716545
|
Bel-Art™
F36730-0021 |
1.6, 3 mL | 21 cm | Fluo-Kem Teflon FEP coated | N/A | ||||||
Description
Double-ended with two bowl sizes.
- Made from 304 stainless-steel
- Sampling spoons have solid, rounded shaft
- Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Spoon | |
Stainless Steel | |
Autoclavable | |
Solid, Rounded Shaft |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bel-Art™ Stainless Steel Spoons