Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Micrometer Pore with ASTM E 11
Brand: RETSCH 60.132.000300
Specifications
1 Each | |
Test Sieve | |
Shaker AS 200 | |
50mm | |
200 mm | |
Stainless Steel |
300 μm | |
ASTM E 11 | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |