Learn More
Bochem™ Stainless Steel Test Tube Stand, Detachable
Transport your test tubes safely with the Bochem™ Stainless Steel Test Tube Stand.
Brand: Bochem™ 8240
This item is not returnable.
View return policy
Description
- Rectangular
- Height: 90mm
Specifications
Test Tube Rack | |
Stainless Steel (18/10) | |
145 x 52mm | |
52 mm | |
17 mm |
Tubes (17 mm) | |
12 | |
145 mm | |
90 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Product Title
Bochem™ Stainless Steel Test Tube Stand, Detachable > Height: 90mm
Spot an opportunity for improvement?Share a Content Correction