Learn More
Epredia™ StainMate™ Mini 10 Batch Stainer Accessories
Maintain Epredia™ StainMate™ Mini 10 Batch Stainer with StainMate Mini 10 Batch Stainer Accessories.
40.10 EUR - 53.95 EUR
Products 3
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12648446
View Documents
Epredia™
E103/SS/BL
Lid N/A N/A N/A
12688446
View Documents
Epredia™
E103/SS/WB
Water dish/bath N/A N/A N/A
12638446
View Documents
Epredia™
E103/SS/B
Additional dish/bath N/A N/A N/A
Description
Description
Accessories for the Mini 10 Batch Stainer Module include:
  • Additional dish/bath
  • Lid
  • Drying module
  • External dish/bath
  • Additional rack
  • Fume hood
  • Bath dish
Dryer Module
  • Fan-assisted slide dryer operates at approx. 70°C
  • Hooks onto either the left- or right-hand side of the StainMate module
Fume Hood
  • Extracts fumes through a replaceable charcoal filter
  • Fits easily inside the base of the hood
  • Transparent door is hinged for easy access
External Dish/Bath
  • Attaches to either side of the StainMate to increase the capacity
  • Constructed of polished stainless steel with white acetal bath and lid
  • Capacity: 38mL
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ StainMate™ Mini 10 Batch Stainer Accessories