missing translation for 'onlineSavingsMsg'
Learn More
Stannous Chloride, 99%, (Crystalline), MP Biomedicals Product Code.: 11426612

Stannous Chloride, 99%, (Crystalline), MP Biomedicals

Product Code. 11426612 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11426612

Brand: MP Biomedicals 0215015791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99%
Formula Weight 189.6
Chemical Name or Material Stannous Chloride, Anhydrous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Stannous Chloride, 99%, (Crystalline), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.