Learn More
Cytiva Steel Spanner Wrenches
Brand:  Cytiva 28-9647-78
 View more versions of this product
Product Code. 12465670
  • 99.00 EUR / Each
In Stock
Specifications
Spanner Wrench
95 to 100 μL
Locking Nuts Diameter 95 to 100 mm
Steel with galvanized surface
1/Pk.
Documents
One moment while we fetch your results.
Product Title
Cytiva Steel Spanner Wrenches > For nuts diameter: 95 to 100mm