Learn More
Bochem™ Steel Zincked Triangles
Made from zincked steel. The Bochem Steel Zincked Triangles come with clay tubes.
2.50 EUR - 2.80 EUR
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10385761
|
Bochem™
12701 |
50mm |
2.50 EUR
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
10652692
|
Bochem™
12702 |
60mm |
2.55 EUR
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10070821
|
Bochem™
12703 |
70mm |
2.80 EUR
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
Description
- Triangles
- Steel zincked
- With clay tubes
Specifications
Steel Zincked | |
Triangle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ Steel Zincked Triangles