Learn More
Gosselin™ Sterile Spoons
Suitable for general laboratory use. Gosselin™ Sterile Spoons are manufactured from sterile polystyrene.
Brand: Gosselin™ CUIG-02
This item is not returnable.
View return policy
Description
- Individually packaged
- Available in two different sizes
Specifications
8 mL | |
White | |
EC N°1935/2004, ISO 11137 |
Product Content Correction
Product Title
Gosselin™ Sterile Spoons > Non-sterile; Capacity: 8mL
Spot an opportunity for improvement?Share a Content Correction