Learn More
Bel-Art™ Sterileware™ Teaspoon Style Sampling Spoon
Spoon is designed for quick and efficient surface sampling of powders and granules. Used by pharmaceutical, food, cosmetic and chemical companies.
Brand:  Bel-Art™ F36941-0010
 View more versions of this product
Product Code. 16692814
  • 125.31 EUR / Pack of 100
Estimated Shipment: 04-07-2025
Description
  • Long handle reaches into jars, bottles and other confined areas
  • Can also be used for dispensing small quantities
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) spoons heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
Spoon
Not Autoclavable
Sterile
Bowl: 4.8 x 3.5 x 1 cm (1.875 x 1.375 x 0.375 in.)
13.7 cm
High-impact Polystyrene
10 mL
100/Pk.
5.375 in.
Product Title
Bel-Art™ Sterileware™ Teaspoon Style Sampling Spoon > 10 mL