Learn More
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
123.00 EUR - 296.00 EUR
Specifications
Product Type | Disposable Scoop |
---|---|
Material | Polystyrene |
Includes | Individually Bagged |
Disposable | Single-use |
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
12917945
![]() |
Sampling Systems™
8045R-60S |
60 mL | Sterile |
146.00 EUR
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
17488873
![]() |
Sampling Systems™
8045R-2500S |
2.5 L | Sterile |
140.00 EUR
Pack of 15 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
15510904
![]() |
Sampling Systems™
8045R-30S |
30 mL | Sterile |
123.00 EUR
Pack of 100 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17418883
![]() |
Sampling Systems™
8045R-500S |
500 mL | Sterile |
296.00 EUR
Pack of 50 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17478873
![]() |
Sampling Systems™
8045R-1000S |
1000 mL | Sterile |
194.00 EUR
Pack of 25 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
15819441
![]() |
Sampling Systems™
8045R-125S |
125 mL | Sterile |
213.00 EUR
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
17498873
![]() |
Sampling Systems™
8045R-250S |
250 mL | Sterile |
213.00 EUR
Pack of 85 |
Estimated Shipment: 09-06-2025
Log in to see stock. |
|||||
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Specifications
Disposable Scoop | |
Individually Bagged | |
Sterile |
Polystyrene | |
Single-use | |
Red |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™