Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Resin
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1 set
1000mL
Description
- Strep-Tactin™XT 4Flow™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
- The low concentrated agarose allows for the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
- In addition, Strep-Tactin™XT 4Flow™ enables the application of denaturing agents such as urea.
- The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
- To pack a column with 1 mL resin bed volume, 2 mL 50% suspension are necessary.
Specifications
Specifications
| Description | High capacity resin |
| Quantity | 1000 mL |
| For Use With (Application) | Purify Strep-Tag™II or Twin-Strep-Tag™ Fusion Proteins |
For Research Use Only.