Learn More
WTW™ Sulfite Reagent Test Kit
With certificate for optimum precision.
Brand: WTW™ 252053
215.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Chromosulfuric Acid oxidation | |
For PhotoLab™ S12, PhotoLab™ Spectral photometers | |
150 | |
Photometric |
For waste water, boiler water, foodstuffs | |
Certificates | |
1.0 - 60.0mg/I SO₃ |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
WTW™ Sulfite Reagent Test Kit > Reagent Test; Detection Range: 1.0 to 60mg/L
Spot an opportunity for improvement?Share a Content Correction