missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur, MP Biomedicals™
Click to view available options
Quantity:
50 g
Description
Pale yellow crystalline solid. Applications include chromatography, analytical and environmental standards, materials science, and pesticides.
Specifications
Specifications
CAS | 7704-34-9 |
Quantity | 50 g |
Molecular Formula | S |
MDL Number | MFCD00085316 |
Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
SMILES | [S] |
IUPAC Name | sulfur |
Molecular Weight (g/mol) | 32.06 |
PubChem CID | 5362487 |
Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'