missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This volumetric solution was checked by means of potentiometric methods using Scharlau's tris(hydroxymethyl)- aminomethane volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
- Molecular Formula: H2SO4
- Molar Mass: 98.08g/mol
- CAS: 7664-93-9
- EIN ECS No.: 231-639-5
- Factor: 0.999–1.001
Specifications
Specifications
CAS | 7664-93-9 |
Concentration | 0.05mol/L |
Density | ∼1.00g/cm3 |
Traceability to NIST | Traceable to NIST |
Normality | 0.1N |
Quantity | 1 L |
Molecular Formula | H2O4S |
Physical Form | Solution |
MDL Number | MFCD00064589 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfuric Acid, 0.05M/l Solution, Scharlab >
Spot an opportunity for improvement?Share a Content Correction