missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, 95–97%, Reagent Grade, Scharlab Product Code.: 11382545
compliance-icons

Sulfuric Acid, 95–97%, Reagent Grade, Scharlab

Product Code. 11382545
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11382545

Brand: Scharlab AC20691000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Used in analytical chemistry, as a laboratory reagent, as an acidifying agent, in the synthesis of organic products and for nitrogen determinations.

  • Molecular Formula: H2SO4
  • Molar Mass: 98.08g/mol
  • CAS: 7664-93-9li>
  • EIN ECS No.: 231-639-5

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point ∼-15°C
Boiling Point ∼310°C
Density 1.84g/cm3
Residue after Ignition 0.0003% max
Vapor Pressure ∼0.0001hPa at 20°C
Quantity 1 L
Physical Form Solution
Solubility Information Miscible in water (20°C)
Formula Weight 98.08g/mol
Percent Purity 95 to 97%
Grade Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid, 95-97%, reagent grade, ISO
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, 95–97%, Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.