missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid, TraceSELECT™, for trace analysis,≥95%, Honeywell Fluka™ Product Code.: 15647320

Sulfuric acid, TraceSELECT™, for trace analysis,≥95%, Honeywell Fluka™

Product Code. 15647320 Shop All Honeywell Fluka Products
2.5 L, Plastic Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Plastic Bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15647320

Brand: Honeywell-Fluka 847162.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

TraceSELECT™, for trace analysis,≥95%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 310°C
CAS Min % ≥50.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%
Density 1.840 g/mL at 25 °C(lit.)
Packaging Plastic Bottle
Quantity 2.5 L
Linear Formula H2SO4
UN Number UN1830
Beilstein 2037554
Formula Weight 98.08
Percent Purity ≥95%
Chemical Name or Material Sulfuric acid, TraceSELECT™, for trace analysis,≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, TraceSELECT™, for trace analysis,≥95%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.