Learn More
Invitrogen™ SureLock™ Tandem Midi Blot Module Replacement Gasket Kit
This kit provides replacement gaskets for the SureLock Tandem Midi Blot Module.
Brand: Invitrogen™ STM2003
Description
About the SureLock Tandem Midi Blot Module
The SureLock Tandem Midi Blot Module is a wet transfer device used exclusively with the SureLock Tandem Midi Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total. The SureLock Tandem Midi Blot Module is compatible with all Invitrogen midi gels (blot size: 8.5 cm x 14 cm). The blot module accommodates one midi gel per wet transfer. The unique design make the module easy to use, while the efficient size of the inner core uses less transfer buffer per transfer (only ∼300 mL per transfer). Since transfer buffer contains methanol, this also results in less hazardous waste requiring special disposal. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 30 minutes and at room temperature.

Specifications
SureLock Tandem Midi Blot Module | |
SureLock | |
SureLock Tandem Midi Blot Module (STM2001) | |
Room Temperature |
Midi | |
Replacement Gasket Kit | |
1 kit | |
1 year |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.