Learn More
Anritsu™ Surface Probe
Brand:  Anritsu™ CS-23K-005-1-TC1-ANP
 View more versions of this product
Product Code. 11972149
  • 189.00 EUR / Each
Estimated Shipment: 27-05-2025
Specifications
6 mm dia.
Product Title
Anritsu™ Surface Probe > Diameter: 6mm