Learn More
Syntaxin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA11042
Product Code. 11505892
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
PA1-1042 detects Syntaxin 1 in rat, mouse and bovine samples. PA1-1042 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ∽33 kDa protein representing Syntaxin 1 in rat, mouse and bovine brain tissue samples. The PA1-1042 immunogen is a synthetic peptide corresponding to residues V(22) T V D R D R F M D E F F E Q V E E I R(41) of rat Syntaxin 1. This sequence is conserved in mouse, human and bovine. The PA1-1042 immunizing peptide (Cat. No. PEP-278) is available for use in neutralization and control experiments.
Syntaxin 1 is a small integral membrane protein that is abundantly expressed in neurons and neuroendocrine cells and thought to be implicated in the regulation of synaptic neurotransmitter release. Syntaxin 1 interacts with several other proteins important for synaptic function (SNAP 25, Munc 18). Syntaxin 1 is the main target of one of the Botulinum neurotoxins (BoNT/C1) which, however, cannot cleave the protein when complexed with its partner proteins in the fusion complex. Syntaxin 1 is an essential component of the neurotransmitter release machinery, and regulation of Syntaxin 1 expression levels is thought to contribute to the mechanism underlying learning and memory.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syntaxin 1
Polyclonal
Unconjugated
STX1A
HPC-1, P35-1, STX1, SYN1A
Rabbit
Antigen affinity chromatography
RUO
116470, 20907, 6804, 788566
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P32850, Q16623, O35526, P32851
STX1A
Synthetic peptide corresponding to residues V(22) T V D R D R F M D E F F E Q V E E I R(41) of rat Syntaxin 1.
100 μg
Primary
Bovine, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Syntaxin 1 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated