Learn More
BD Syringes
Brand:  BD 307740
Product Code. 15532961
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
23 G
25 mm
2000/Pk.
0.6 mm
Syringe
2 mL
Product Title
BD Syringes > Volume: 2mL; With Removable Needle