Learn More
Applied Biosystems™ TaqMan™ Array Human Hypoxia
TaqMan™ Array Human Hypoxia 96-well Plate contains 44 assays to hypoxia signaling associated genes and 4 assays to candidate endogenous control genes.
Brand: Applied Biosystems™ 4414090
156.67 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The panel of assays in this plate targets genes in the HIF (hypoxia inducible factor) family. On the panel are the genes VHL, Cul2, and RBX1 (Ring-Box) that complex with additional genes for a role in the ubiquitylation of HIF1A. Other hypoxia response related genes fall in to the following target classes: Transcription Factors, Immune and Stress Response, Signal Transduction, Apoptosis, Oxidoreductases, Proteases, and Protein metabolism. Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate TaqMan Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.
TaqMan Array Plates contain the same TaqMan Gene Expression Assays that have been available for years in a dried, 96-well format. Tests have found no difference in performance between TaqMan Assays supplied in the liquid format and assays dried into a TaqMan Array Plate. Ideal for projects such as validation of microarray and RNAi data, the plates include enough TaqMan Assay in each well for one 20uL reaction. Each pack of TaqMan Array Plates is shipped with a CD containing the plate map along with details of each TaqMan Assay. This information is imported into the real-time PCR instrument to enable proper labeling of data and easy access to Assay ID numbers for simple reordering.
- Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
- Fast delivery; on your bench in days
- Convenient; dried PCR primer and TaqMan probe sets supplied in a 96- well plate; just add master mix and your sample
- TaqMan Array Plates have been optimized for use with TaqMan Gene Expression Master Mix, but are also compatible with TaqMan Universal PCR Master Mix
- For studies using cultured cells, TaqMan Array Plates are also compatible (and recommended) with the TaqMan Gene Expression Cells-to-Ct™ Kit
- TaqMan Array Plates must be run on a real-time PCR instrument with 96-well plate capability
- The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μL reaction volumes
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
1 standard plate | |
Human | |
1 x 20 μL Reactions |
Dried Down | |
Standard | |
FAM | |
Standard | |
Cell Cycle & Proliferation, Neurobiology, Oncology, Development, Immunology, Signal Transduction | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.