Learn More
Taranabant ((1R,2R)stereoisomer), MedChemExpress
MedChemExpress Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.
Brand:  Medchem Express HY-10013B
Encompass_Preferred
 View more versions of this product
Product Code. 16402798
  • 766.80 EUR / 10mg
Description
Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.
Specifications
Specifications
Taranabant ((1R,2R)stereoisomer)
701977-08-4
10 mg
C27H25ClF3N3O2
DMSO : 100 mg/mL (193.82 mM; Need ultrasonic)
515.95
98.15%
Solid
Off-White
Research
MK0364 (1R,2R)stereoisomer
CC(OC1=NC=C(C(F)(F)F)C=C1)(C)C(N[C@H](C)[C@@H](C2=CC=CC(C#N)=C2)CC3=CC=C(Cl)C=C3)=O
515.95
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Taranabant ((1R,2R)stereoisomer), MedChemExpress