Learn More
Taranabant ((1R,2R)stereoisomer), MedChemExpress
MedChemExpress Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.
Brand: Medchem Express HY-10013B
Description
Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.Specifications
Taranabant ((1R,2R)stereoisomer) | |
701977-08-4 | |
10 mg | |
C27H25ClF3N3O2 | |
DMSO : 100 mg/mL (193.82 mM; Need ultrasonic) | |
515.95 | |
98.15% |
Solid | |
Off-White | |
Research | |
MK0364 (1R,2R)stereoisomer | |
CC(OC1=NC=C(C(F)(F)F)C=C1)(C)C(N[C@H](C)[C@@H](C2=CC=CC(C#N)=C2)CC3=CC=C(Cl)C=C3)=O | |
515.95 | |
Research |
Product Suggestions
Customers who viewed this item also viewed