Learn More
TCS 401, MedChemExpress
MedChemExpress TCS 401 is a selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).
Brand: Medchem Express HY-12312
Description
TCS 401 is a selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).Specifications
TCS 401 | |
243966-09-8 | |
10 mg | |
C10H11ClN2O5S | |
0.1 M NaOH : 6.67 mg/mL (21.75 mM; ultrasonic and adjust pH to 9 with NaOH) | |
306.72 | |
98.0% |
Solid | |
Earth Yellow | |
Research | |
Metabolism-sugar/lipid metabolism | |
O=C(C1=C(NC(C(O)=O)=O)SC2=C1CCNC2)O.[H]Cl | |
306.72 | |
Research |