missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Assay Percent Range | 94% min. (GC) |
Quantity | 1 g |
Formula Weight | 200.28 |
Percent Purity | 95% |
Chemical Name or Material | tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate |
Safety and Handling

Product Identifier
- tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction