Learn More
Memmert™ Test Tube Rack
Test Tube Rack designed for use with a Memmert™ Water Bath.
Brand: Memmert™ WTB35B50656
This item is not returnable.
View return policy
Description
- Water Bath Accessory
Specifications
WTB35 Water Baths | |
74 Test Tubes (2 Rows) | |
10 mm |
Test Tube Rack | |
Stainless Steel |
Product Content Correction
Product Title
Memmert™ Test Tube Rack > For Use With: WTB35 Water Baths; Holds: 74 Test Tubes (2 Rows); Tube Diameter: 10 mm
Spot an opportunity for improvement?Share a Content Correction