missing translation for 'onlineSavingsMsg'
Learn More
Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals Product Code.: 11358367

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Product Code. 11358367
4 L
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 11358367

Brand: Thermo Scientific Alfa Aesar 032437.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Tetrachloroethylene is used as solvent. It is involved in dry cleaning as well as in the degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostinShow MoreShow Less
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name 1,1,2,2-tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point °C to 22°C
Density 1.623 g/mL
Boiling Point 120°C to 121°C
Assay Percent Range >99%
Refractive Index 1.5055
Quantity 4 L
Beilstein 1361721
Solubility Information Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water.
Formula Weight 165.83
Grade Spectrophotometric
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene, Ultrapure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ05U901 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number032437.K2,032437.M1,032437.K7
Lot NumberJ20W896 Certificate TypeCertificate of Analysis Date30/06/2022 Catalog Number032437.K2,032437.M1,032437.K7
Lot NumberH12Z705 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number032437.K2,032437.M1,032437.K7
Lot NumberY11C700 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number032437.K2,032437.M1,032437.K7
Lot NumberC23W044 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number032437.K2,032437.M1,032437.K7
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrachloroethylene, Ultrapure, Spectrophotometric Grade
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tetrachloroethylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.