missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran, BHT Stabilized, Multisolvent™ Grade, Sharlab Product Code.: 11342575

Tetrahydrofuran, BHT Stabilized, Multisolvent™ Grade, Sharlab

Product Code. 11342575
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 11342575

Brand: Scharlab TE02281000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used for almost every laboratory application. Tetrahydrofuran, BHT Stabilized, Multisolvent™ Grade from Sharlab features high purity and low water content for use in applications requiring an ACS-grade, GPC-grade or Reagent-grade product.

Multisolvent™ products are most frequently use for HPLC, but can also be used for numerous other applications, making it one of the most universal grades available. These versatile products have numerous advantages over application-specific solvents:

  • Guarantee results: QC controls are performed by modern analytical techniques for all guaranteed applications
  • Optimize purchasing and stocking: One product for most applications prevents the need to order, stock and track the supply of multiple grade
  • Optimal packaging: Highly-resistant and durable amber glass bottles or stainless steel drums ensure the highest product safety and quality
  • Lower costs by unifying: Multiple-use products at single-grade prices to avoid buying small quantities of specific products
  • Preserve the environment: Returnable packaging (one-way glass bottles or SS returnable drums) help prevent waste

Specifications

Melting Point -108.5°C
CAS 109-99-9
Color Colorless
Acidity Max. 0.0003meq/g
Alkalinity Max. 0.0002meq/g
Appearance Clear
Density 0.89g/cm3
Boiling Point 65°C to 66°C
Quantity 1 L
Flash Point −21.5°C
Identification Passes test
Ignition Point 215°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.025%
Molecular Formula C4H8O
MDL Number MFCD00005356
Vapor Pressure 173 hPa
UN Number 2056
Water Max. 0.02%
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
Solubility Information Miscible in water
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11g/mol
Percent Purity Min. 99.9%
Grade Multisolvent™
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tetrahydrofurian
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : tetrahydrofurian
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran, BHT Stabilized, Multisolvent™ Grade, Sharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.