missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA572495
338.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
TGM4 is associated with the mammalian reproductive process. TGM4 catalyzes the cross-linking of proteins and the conjugation of polyamines to specific proteins in the seminal tract.
Specifications
TGM4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P49221 | |
TGM4 | |
KLH conjugated synthetic peptide between 135-164 amino acids from the Central region of human TGM4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TGM4 | |
9530008N10Rik; Dorsal prostate transglutaminase; dorsal protein 1; DP1; Eapa1; Experimental autoimmune prostatitis antigen 1; fibrinoligase; FLJ26776; hTGP; prostate transglutaminase; Prostate-specific transglutaminase; protein-glutamine gamma-glutamyltransferase 4; TG; TG(P); TGase P; TGase TGase-4; TGase-4; TGM4; TGP; transglutaminase 4; transglutaminase 4 (prostate); transglutaminase P; Transglutaminase-4 | |
Rabbit | |
Protein A | |
RUO | |
7047 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGM4 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction