Learn More
Abeba™ The Original 7020 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
56.85 EUR - 59.50 EUR
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Style | 7020 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15542086
|
Abeba™
7020/35 |
35 |
56.85 EUR
1 pair |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||||
15582086
|
Abeba™
7020/36 |
36 |
59.50 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15512096
|
Abeba™
7020/37 |
37 |
56.85 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15532096
|
Abeba™
7020/38 |
38 |
59.45 EUR
1 pair |
Estimated Shipment: 15-04-2025
Log in to see stock. |
||||||
15552096
|
Abeba™
7020/39 |
39 |
56.85 EUR
1 pair |
Estimated Shipment: 15-04-2025
Log in to see stock. |
||||||
15562096
|
Abeba™
7020/40 |
40 |
56.85 EUR
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15572096
|
Abeba™
7020/41 |
41 |
56.85 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15582096
|
Abeba™
7020/42 |
42 |
57.75 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15502106
|
Abeba™
7020/43 |
43 |
56.85 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15512146
|
Abeba™
7020/44 |
44 |
56.85 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15522146
|
Abeba™
7020/45 |
45 |
57.75 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15552146
|
Abeba™
7020/46 |
46 |
56.85 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15562146
|
Abeba™
7020/47 |
47 |
57.75 EUR
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
Description
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather, perforated
- White
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB FO SRC | |
Leather | |
7020 |
White | |
Slip-resistant | |
Animal fats, oil and fuel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Abeba™ The Original 7020 Shoes