Learn More
Julabo™ Thermal Bath Fluids
Used for temperature control applications. Julabo™ Thermal Bath Fluids offer safe and reliable operation.
Brand: Julabo™ 8940106
This item is not returnable.
View return policy
Description
- For closed systems like the Presto or Forte
- Has wide temperature ranges
- Low viscosity
- High stability
- Good heat conductivity
- Minimum odor
- Low corrosion tendency
- Low toxicity
- Long shelf life
Specifications
Clear | |
-50°C to +105°C | |
Thermal Bath | |
H5 | |
>400°C |
Refrigerated circulators | |
Thermal Bath Fluid | |
Drain tap | |
10 L | |
5.66mm2/s at 20°C |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Julabo™ Thermal Bath Fluids > Color: Clear; Temp Range: -50 to 105 deg.C; Fire Point: 142 deg.C; Qty.: 10L
Spot an opportunity for improvement?Share a Content Correction