Learn More
Epredia™ Shandon™ Plastic Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Brand:  Epredia™ 72110017
153.00 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 12766807
  • 180.09 EUR / Pack of 25
Description
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coverplate
25/Pk.
Product Title
Epredia™ Shandon™ Plastic Coverplates > Coverplate; 25 Pk.