Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
Shop All Thermo Scientific Products
Click to view available options
Color:
Brown
Gray
Green
Green/Yellow
Red
White
Unit Size:
Each
Description
- Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifications
Specifications
| For Use With (Equipment) | 4 x 180 Swing bucket rotor |
| Color | Green/Yellow |
Product Content Correction
Product Title
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor >
Spot an opportunity for improvement?Share a Content Correction