Learn More
Testo™ Thermocouple
Brand: Testo™ 0602 0644
This item is not returnable.
View return policy
Specifications
Thermocouple, 1.5 dia. x 800mm L Probe Shaft, Fibre Glass, –50 to 400°C Measuring Range, 5 sec. Reaction Time, Flexible, With TC Adapter | |
EN 60584-2 | |
Fiberglass | |
5 sec. |
Product Content Correction
Product Title
Testo™ Thermocouple > Probe Diameter: 1.5mm
Spot an opportunity for improvement?Share a Content Correction