missing translation for 'onlineSavingsMsg'
Learn More
Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals Product Code.: 11423930

Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 11423930
500 g
Click to view available options
Quantity:
100 g
500 g
1000 g
This item is not returnable. View return policy

Product Code. 11423930

Brand: Thermo Scientific Alfa Aesar A14344.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

Solubility
Miscible with chloroform, alcohol, ether, dimethylsulfoxide, ethyl acetate and most organic solvents. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-03-3
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.15
MDL Number MFCD00005429
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiopheneShow MoreShow Less
PubChem CID 7364
ChEBI CHEBI:87301
IUPAC Name thiophene-2-carbaldehyde
SMILES O=CC1=CC=CS1

Specifications

Density 1.22
Boiling Point 196°C to 198°C
Flash Point 78°C (172°F)
Refractive Index 1.591
Quantity 500 g
Beilstein 105819
Sensitivity Air sensitive
Solubility Information Miscible with chloroform,alcohol,ether,dimethylsulfoxide,ethyl acetate and most organic solvents. Immiscible with water.
Formula Weight 112.15
Percent Purity ≥98%
Chemical Name or Material Thiophene-2-carboxaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251183 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14344.22,A14344.36,A14344.0B
Lot Number10226771 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog NumberA14344.22,A14344.36,A14344.0B
Lot Number10223359 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog NumberA14344.22,A14344.36,A14344.0B
Lot Number10243093 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog NumberA14344.22,A14344.36,A14344.0B
Lot Number10109935 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14344.22,A14344.36,A14344.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.