Learn More
Thiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Description
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.

Specifications
Specifications
Melting Point | 29°C to 31°C |
Boiling Point | 80°C to 81°C (1 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.573 |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 123536 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 182.65 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10244482 | Certificate of Analysis | 09/11/2022 | A15553.06,A15553.14 |
10237802 | Certificate of Analysis | 12/04/2022 | A15553.06,A15553.14 |
10188730 | Certificate of Analysis | 23/11/2021 | A15553.06,A15553.14 |
E1377C | Certificate of Analysis | 23/11/2021 | A15553.06,A15553.14 |
10177151 | Certificate of Analysis | 23/11/2021 | A15553.06,A15553.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.