Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand: SymLine™ 106475
224.49 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Angled Shut-off Adapter, 1 x GL25 (f) to 1 x GL25 (m) | |
Black | |
HDPE-EC | |
Adapter |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
37Hours
20Minutes
55Seconds
56New Offer Save Up to 70%